Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(3-amino-4-methylphenyl)hexafluoropropane 98.0+%, TCI America™
SDP

Catalog No. B13975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13975G Supplier TCI America Supplier No. B13975G

Specifications

CAS 116325-74-7
Melting Point 115°C
Molecular Formula C17H16F6N2
Quantity 5 g
MDL Number MFCD00039148
Synonym 5,5′C-(Hexafluoroisopropylidene)di-o-toluidine
InChI Key HJSYPLCSZPEDCQ-UHFFFAOYSA-N
SMILES CC1=C(N)C=C(C=C1)C(C1=CC(N)=C(C)C=C1)(C(F)(F)F)C(F)(F)F
IUPAC Name 5-[2-(3-amino-4-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-methylaniline
Molecular Weight (g/mol) 362.32
PubChem CID 632384
Formula Weight 362.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(3-amino-4-methylphenyl)hexafluoropropane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.