Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(3-aminophenyl)hexafluoropropane 98.0+%, TCI America™
SDP

Catalog No. B17045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17045G Supplier TCI America Supplier No. B17045G

Chemical Identifiers

CAS 47250-53-3
Molecular Formula C15H12F6N2
Molecular Weight (g/mol) 334.265
MDL Number MFCD00191619
InChI Key UVUCUHVQYAPMEU-UHFFFAOYSA-N
Synonym 3,3′-(Hexafluoroisopropylidene)dianiline
PubChem CID 630566
IUPAC Name 3-[2-(3-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
SMILES C1=CC(=CC(=C1)N)C(C2=CC(=CC=C2)N)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 86°C
Quantity 5 g
Formula Weight 334.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(3-aminophenyl)hexafluoropropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.