Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane 99.0+%, TCI America™
SDP

Catalog No. B202125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B202125G 25 g
B2021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B202125G Supplier TCI America Supplier No. B202125G

Chemical Identifiers

CAS 25327-89-3
Molecular Formula C21H20Br4O2
Molecular Weight (g/mol) 624.005
MDL Number MFCD00086863
InChI Key PWXTUWQHMIFLKL-UHFFFAOYSA-N
Synonym Tetrabromobisphenol A Diallyl Ether
PubChem CID 61972
IUPAC Name 1,3-dibromo-5-[2-(3,5-dibromo-4-prop-2-enoxyphenyl)propan-2-yl]-2-prop-2-enoxybenzene
SMILES CC(C)(C1=CC(=C(C(=C1)Br)OCC=C)Br)C2=CC(=C(C(=C2)Br)OCC=C)Br

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 25 g
Formula Weight 624.01
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane
EINECSNumber (4)-0218
RTECSNumber DA6126250
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.