Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene 99.0+%, TCI America™
SDP

Catalog No. B01335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B01335G 5 g
B01331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B01335G Supplier TCI America Supplier No. B01335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 72-55-9
Molecular Formula C14H8Cl4
Molecular Weight (g/mol) 318.02
MDL Number MFCD00000837
InChI Key UCNVFOCBFJOQAL-UHFFFAOYSA-N
Synonym p,p'-dde, dichlorodiphenyldichloroethylene, ddt dehydrochloride, 4,4'-dde, 2,2-bis 4-chlorophenyl-1,1-dichloroethylene, dde, 1,1-dichloro-2,2-bis 4-chlorophenyl ethene, p,p'-dichlorodiphenyldichloroethylene, p,p'-dichlorodiphenyl dichloroethylene, p,p'-dichlorodiphenyl-2,2-dichloroethylene
PubChem CID 3035
ChEBI CHEBI:16598
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
SMILES ClC(Cl)=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 89°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 318.02
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene
RTECSNumber KV9450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.