Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(4-hydroxyphenyl)butane 98.0+%, TCI America™
SDP

Catalog No. B10505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10505G 5 g
B105025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B10505G Supplier TCI America Supplier No. B10505G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77-40-7
Molecular Formula C16H18O2
Molecular Weight (g/mol) 242.32
MDL Number MFCD00020179
InChI Key HTVITOHKHWFJKO-UHFFFAOYSA-N
Synonym bisphenol b, 2,2-bis 4-hydroxyphenyl butane, 4,4'-butane-2,2-diyl diphenol, phenol, 4,4'-1-methylpropylidene bis, p,p'-sec-butylidenediphenol, unii-1rc731tjja, 4,4'-1-methylpropylidene bisphenol, 1rc731tjja, 4,4'-sec-butylidenediphenol, 2,2-bis p-hydroxyphenyl butane
PubChem CID 66166
ChEBI CHEBI:34581
IUPAC Name 4-[2-(4-hydroxyphenyl)butan-2-yl]phenol
SMILES CCC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 5 g
Formula Weight 242.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-hydroxyphenyl)butane
RTECSNumber SM1210200
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.