Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2-Bis(aminoethoxy)propane

Catalog No. 7732041G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-320-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-320-41G Supplier Sigma Aldrich Supplier No. 7732041G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2-Bis(aminoethoxy)propane is an acid degradable amine monomer that can be used as a ketal containing cross-linker. It is a pH sensitive molecule that is used in the synthesis of polyurea and polyurethane.This homobifunctionalized amino ketal is primarily used in crosslinking cell lysate (linking polypeptides whose lysine side chains are nearest to each other). Primary amines commonly conjugate through acylation or alkylation of isothiocyanates; isocyanates; acyl azides; NHS esters; sulfonyl chlorides; aldehydes; carbonates; aryl halides; imidoesters; carbodiimides; anhydrides; and fluorophenyl esters. This diamine can also crosslink epoxies.

Specifications

CAS 127090-71-5
Density 1.018 g/mL (at 25°C)
Boiling Point 202°C to 214 (760 mmHg)
Molecular Formula C7H18N2O2
Linear Formula C7H18N2O2
Refractive Index n20/D 1.458
MDL Number MFCD20694614
Quantity 1 g
Synonym 2,2 -[(1-Methylethylidene)bis(oxy)]bis(ethanamine); 2,2-Di(2-aminoethoxy)propane; Diamino ketal
Molecular Weight (g/mol) 162.23
Percent Purity ≥98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.