Learn More
Strem, An Ascensus Company CAS# 84783-64-2. 250mg. 2,2'-Bis(diphenylphosphino)-1,1'-biphenyl, 98% BIPHEP. MFCD03094574
SDP
Catalog No. 5090109788
Encompass
CAS# 84783-64-2. 250mg. 2,2'-Bis(diphenylphosphino)-1,1'-biphenyl, 98% BIPHEP. MFCD03094574. Molecular Weight: 522.57. Molecular Formula: C36H28P2. Color/form: white xtl. Strem# 15-0145. http://www.strem.com/catalog/v/15-0145/
Catalog No. 50-901-09788 Supplier Strem, An Ascensus Company Supplier No. 150145250MG
CAS# 84783-64-2. 250mg. 2,2'-Bis(diphenylphosphino)-1,1'-biphenyl, 98% BIPHEP. MFCD03094574. Molecular Weight: 522.57. Molecular Formula: C36H28P2. Color/form: white xtl. Strem# 15-0145. http://www.strem.com/catalog/v/15-0145/