Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97+%

Catalog No. AAH2622622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2622622 100 g
AAH2622606 5 g
AAH2622614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2622622 Supplier Thermo Scientific Chemicals Supplier No. H2622622
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 98327-87-8 | C44H32P2 | 622.69 g/mol

(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl plays an essential role in the organic synthesis of enantioselective transformations catalyzed by the complexes of ruthenium, rhodium and palladium. It is also employed in palladium-catalyzed arylamine coupling in the preparation of demethylthiocholchines. It is used with Cu(II) to catalyze the addition of arylsulfonamides to styrenes and olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl plays an essential role in the organic synthesis of enantioselective transformations catalyzed by the complexes of ruthenium, rhodium and palladium. It is also employed in palladium-catalyzed arylamine coupling in the preparation of demethylthiocholchines. It is used with Cu(II) to catalyze the addition of arylsulfonamides to styrenes and olefins.

Solubility
Soluble in tetrahydrofuran, benzene and dichloromethane. Slightly soluble in ether, methanol and ethanol. Insoluble in water.

Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98327-87-8
Molecular Formula C44H32P2
Molecular Weight (g/mol) 622.69
MDL Number MFCD00010805
InChI Key MUALRAIOVNYAIW-UHFFFAOYSA-N
Synonym binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthalene
PubChem CID 634876
IUPAC Name [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=C(C2=CC=CC=C2C=C1)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 281°C to 285°C
Quantity 100 g
Beilstein 5321443
Merck Index 14,1223
Solubility Information Soluble in tetrahydrofuran,benzene and dichloromethane. Slightly soluble in ether,methanol and ethanol. Insoluble in water.
Formula Weight 622.69
Percent Purity 98%
Chemical Name or Material (+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 619-338-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.