Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%

Catalog No. AAB2251536
missing translation for 'changeView'
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2251536 500 g
AAB2251514 25 g
AAB2251522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AAB2251536 Supplier Thermo Scientific Chemicals missing translation for 'supplierNo' B2251536
Only null left
Add to cart

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol

It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

Solubility
Soluble in water (209.2 g/L), at 20°C and alcohol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6976-37-0
Molecular Formula C8H19NO5
Molecular Weight (g/mol) 209.242
MDL Number MFCD00002853
InChI Key OWMVSZAMULFTJU-UHFFFAOYSA-N
Synonym bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane
PubChem CID 81462
ChEBI CHEBI:41250
IUPAC Name 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CO)N(CCO)C(CO)(CO)CO

Specifications

Melting Point 102°C to 105°C
Linear Formula (HOCH2)3CN(CH2CH2OH)2
Quantity 500 g
Beilstein 2205275
Solubility Information Soluble in water (209.2g/L),at 20°C and alcohol.
Formula Weight 209.24
Percent Purity ≥98%
Chemical Name or Material 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P390-P501c
EINECSNumber 230-237-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.