Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Bithiophene-5-boronic acid pinacol ester

Catalog No. 5788781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-887-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-887-81G Supplier Sigma Aldrich Supplier No. 5788781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2'-Bithiophene-5-boronic acid pinacol ester

Specifications

CAS 479719-88-5
Melting Point 35.5°C to 38.0°C (average)
Molecular Formula C14H17BO2S2
Linear Formula C14H17BO2S2
Refractive Index n20/D 1.5900 (literature)
Quantity 1 g
MDL Number MFCD04039974
Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2 -bithiophene
Molecular Weight (g/mol) 292.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.