Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bithiophene-5-boronic acid pinacol ester, 98%

Catalog No. p-7052620
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3798903 1 g
AAH3798906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3798903 Supplier Thermo Scientific Chemicals Supplier No. H3798903
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 479719-88-5 | C14H17BO2S2 | 292.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479719-88-5
Molecular Formula C14H17BO2S2
Molecular Weight (g/mol) 292.22
MDL Number MFCD04039974
InChI Key HPOQARMSOPOZMW-UHFFFAOYSA-N
Synonym 2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester
PubChem CID 3592790
IUPAC Name 4,4,5,5-tetramethyl-2-(5-thiophen-2-ylthiophen-2-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1

Specifications

Melting Point 35°C to 38°C
Color Blue-Green to White
Quantity 1 g
Formula Weight 292.2
Percent Purity 98%
Physical Form Fused Solid
Chemical Name or Material 2,2'-Bithiophene-5-boronic acid pinacol ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.