Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-[2-(Boc-amino)ethoxy]ethanol

Catalog No. 7496485G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-964-85G 5 g
74-964-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-964-85G Supplier Sigma Aldrich Supplier No. 7496485G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-[2-(boc-amino)ethoxy]ethanol is n-terminal protected 2-(2-aminoethoxy)ethanol. it is generally used as a molecular spacer/linker. some of such applications are:
  • Synthesis of biotinylated haloligands to prepare quantum dot adducts for halotag protein-mediated specific labeling of live cells
  • It is one of the key precursors for the total synthesis of tyroscherin and photo-activatable tyroscherin-based affinity reagent used for identification of tyroscherin binding proteins
  • It can be used to link halotag enzyme with fluorescent zinc indicator, zimir to monitor the dynamics of regulated secretion of zinc in real time
  • It can also be used to synthesize amphiphiles for DNA and sirna delivery

Specifications

CAS 139115-91-6
Density 1.061 g/mL (at 25°C)
Molecular Formula C9H19NO4
Refractive Index n20/D 1.454
Linear Formula C9H19NO4
MDL Number MFCD04038542
Quantity 5 g
Synonym 2-(2-tert-Butoxycarbonylaminoethoxy)ethanol; N-[2-(2-Hydroxyethoxy)ethyl]carbamic acid tert-butyl ester; tert-Butyl[2-(2-hydroxyethoxy)ethyl]carbamate; [2-(2-Hydroxyethoxy)ethyl]carbamic acid 1,1-dimethylethyl ester
Molecular Weight (g/mol) 205.25
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.