Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromophenyl)-1,3-dioxolane 97.0+%, TCI America™
SDP

Catalog No. B20845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20845G 5 g
B208425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20845G Supplier TCI America Supplier No. B20845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34824-58-3
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00155124
InChI Key IWSGKSUCFVOWQU-UHFFFAOYSA-N
Synonym 2-2-bromophenyl-1,3-dioxolane, 2-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-2-bromophenyl, 1-bromo-2-1,3-dioxolan-2-yl benzene, 2-bromophenyldioxolane, 2-bromo benzaldehyde ethylene glycol acetal, 2-2-bromo-phenyl-1,3 dioxolane, pubchem3729, acmc-1csyy, bromobenzaldehyde ethylene acetal
PubChem CID 553534
IUPAC Name 2-(2-bromophenyl)-1,3-dioxolane
SMILES BrC1=CC=CC=C1C1OCCO1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Bromophenyl)-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.