Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Butoxyethoxy) Ethyl Acetate, Spectrum™ Chemical
SDP

Catalog No. 18620219 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Metal
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-620-219 1 L Amber Glass Bottle
18-620-220 4 L Amber Glass Bottle
18-620-221 4 x 4 L Amber Glass Bottle
18-620-222 20 L Metal
18-620-223 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-219 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B11791LTGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 124-17-4
Molecular Weight (g/mol) 204.27

Specifications

Quantity 1 L
Packaging Amber Glass Bottle
Boiling Point 238°C to 250°C
Percent Purity 98%
Grade Ungraded
Chemical Name or Material 2-(2-Butoxyethoxy) Ethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.