Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Butoxyethoxy) Ethyl Acetate, Spectrum™ Chemical
SDP

Catalog No. 18620223 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Metal
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-620-223 200 L Drum
18-620-219 1 L Amber Glass Bottle
18-620-220 4 L Amber Glass Bottle
18-620-221 4 x 4 L Amber Glass Bottle
18-620-222 20 L Metal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-223 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1179200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124-17-4
Molecular Weight (g/mol) 204.27

Specifications

Quantity 200 L
Packaging Drum
Molecular Formula C10H20O4
Boiling Point 238°C to 250°C
InChI Key VXQBJTKSVGFQOL-UHFFFAOYSA-N
SMILES CCCCOCCOCCOC(C)=O
IUPAC Name 2-(2-butoxyethoxy)ethyl acetate
Percent Purity 98%
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.