Learn More
Medchemexpress LLC 2-[(2-carboxyphenyl)amino]-4-chlorobenzoic acid | 63329-53-3 | MFCD00866065 | 99.4% | 291.68 g/mol | C14H10ClNO4 | 50MG
SDP
Catalog No. 5000237302
Encompass_Preferred
Lobenzarit is a small-molecule immunomodulator used in preclinical research that has reported anti-oxidative and anti-inflammatory properties. It is supplied as a high-purity research reagent (CAS 63329-53-3) and is used in vitro and in vivo to study immune modulation and anti-arthritic effects.
  • High purity suitable for research use.
  • Reported immunomodulatory and anti-oxidative activity.
  • Available in multiple small-scale sizes for preclinical studies.
  • Stable when stored under recommended conditions (powder and in solvent).
  • Molecular formula C14H10ClNO4; molecular weight 291.68 g/mol.
Catalog No. 50-002-37302 Supplier Medchemexpress LLC Supplier No. HY12441150MG
Lobenzarit is a small-molecule immunomodulator used in preclinical research that has reported anti-oxidative and anti-inflammatory properties. It is supplied as a high-purity research reagent (CAS 63329-53-3) and is used in vitro and in vivo to study immune modulation and anti-arthritic effects.
  • High purity suitable for research use.
  • Reported immunomodulatory and anti-oxidative activity.
  • Available in multiple small-scale sizes for preclinical studies.
  • Stable when stored under recommended conditions (powder and in solvent).
  • Molecular formula C14H10ClNO4; molecular weight 291.68 g/mol.