Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[[2-chloro-5-(phenylsulfonyl)benzoyl]amino]benzoic acid | 724440-27-1 | 98.8% | 5 MG
SDP

Catalog No. 5000233851
Encompass_Preferred

BAY-8002 is a selective, orally active monocarboxylate transporter 1 (MCT1) inhibitor used in preclinical research. It inhibits MCT1-mediated lactate transport (IC50 ≈ 85 nM) and shows in vivo antitumor activity in mouse models. For research use only.

  • Selective MCT1 inhibitor (IC50 ≈ 85 nM in DLD-1 cells).
  • High purity solid, white to off-white.
  • Molecular weight 415.85; formula C20H14ClNO5S.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Packaged as a 5 mg research-grade sample.

Catalog No. 50-002-33851 Supplier Medchemexpress LLC Supplier No. HY1223125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BAY-8002 is a selective, orally active monocarboxylate transporter 1 (MCT1) inhibitor used in preclinical research. It inhibits MCT1-mediated lactate transport (IC50 ≈ 85 nM) and shows in vivo antitumor activity in mouse models. For research use only.

  • Selective MCT1 inhibitor (IC50 ≈ 85 nM in DLD-1 cells).
  • High purity solid, white to off-white.
  • Molecular weight 415.85; formula C20H14ClNO5S.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Packaged as a 5 mg research-grade sample.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.