Learn More
Medchemexpress LLC 2-[[2-chloro-5-(phenylsulfonyl)benzoyl]amino]benzoic acid | 724440-27-1 | 98.8% | 50 MG
SDP
Catalog No. 5000233787
Encompass_Preferred
BAY-8002 is a selective, orally active inhibitor of monocarboxylate transporter 1 (MCT1) used in preclinical research on cancer metabolism; it inhibits MCT1-mediated lactate transport with a reported IC50 of ≈85 nM.
  • Selective MCT1 inhibitor with reported IC50 ≈ 85 nM.
  • Molecular formula C20H14ClNO5S and molecular weight 415.85.
  • High purity (98.8% by HPLC as reported).
  • Supplied as a powder with recommended storage at -20°C for long-term storage or 4°C for short-term.
  • Used in metabolic and oncology research applications.
Catalog No. 50-002-33787 Supplier Medchemexpress LLC Supplier No. HY12231250MG
BAY-8002 is a selective, orally active inhibitor of monocarboxylate transporter 1 (MCT1) used in preclinical research on cancer metabolism; it inhibits MCT1-mediated lactate transport with a reported IC50 of ≈85 nM.
  • Selective MCT1 inhibitor with reported IC50 ≈ 85 nM.
  • Molecular formula C20H14ClNO5S and molecular weight 415.85.
  • High purity (98.8% by HPLC as reported).
  • Supplied as a powder with recommended storage at -20°C for long-term storage or 4°C for short-term.
  • Used in metabolic and oncology research applications.