Learn More
Medchemexpress LLC 2-(2-chlorophenyl)-2,2-diphenylacetonitrile | 197525-99-8 | 99.7% | 303.8 g/mol | C20H14ClN | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10330810MM 1ML
Encompass_Preferred
TRAM-39 (2-(2-chlorophenyl)-2,2-diphenylacetonitrile; C20H14ClN; MW 303.8 g/mol) is a selective blocker of intermediate-conductance Ca2+-activated K+ (KCa3.1/IKCa) channels used in electrophysiology and ion-channel research. Provided as a 10 mM solution in a 1 mL vial, the compound is characterized by high purity and is intended for in vitro pharmacology and biochemical assays.
  • Selective blocker of KCa3.1 (IKCa) channels for ion-channel studies.
  • Provided as a 10 mM solution in a 1 mL vial for convenient dosing.
  • High purity suitable for biochemical and pharmacological assays.
  • Identified by CAS 197525-99-8 and standard chemical identifiers for traceability.
  • Certificate of analysis and safety data sheet available to support use and handling.
Catalog No. 50-002-47314