Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2-chlorophenyl)-2-(5,7-dichloro-3-(hydroxymethyl)-4,6-dimethyl-1H-indol-2-yl)-N-((4-methylphenyl)su | 2244452-09-1 | 99.6% | 499.36 g/mol | C21H20Cl2N2O6S | 1MG
SDP

Catalog No. 5000236658
Encompass_Preferred

BI-4924 is a potent, selective small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) used in biochemical and cellular research to probe serine biosynthesis and PHGDH-dependent metabolism. The compound is a hydroxymethylene-bearing carboxylic acid derivative (CAS 2244452-09-1) with characterized physicochemical and solubility properties.

  • Potent PHGDH inhibition: IC50 = 3 nM.
  • Disrupts serine biosynthesis in cells (IC50 ≈ 2.2 μM at 72 h).
  • Chemical formula C21H20Cl2N2O6S; molecular weight 499.36 g/mol.
  • High purity: 99.6% (reported by supplier).
  • Form: powder; suitable for DMSO stock solutions.
  • Solubility in vitro: DMSO 125 mg/mL (≈250 mM; ultrasonic recommended).
  • In vivo formulations documented for multiple vehicles at ≥2.08 mg/mL.

Catalog No. 50-002-36658 Supplier Medchemexpress LLC Supplier No. HY1262541MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-4924 is a potent, selective small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) used in biochemical and cellular research to probe serine biosynthesis and PHGDH-dependent metabolism. The compound is a hydroxymethylene-bearing carboxylic acid derivative (CAS 2244452-09-1) with characterized physicochemical and solubility properties.

  • Potent PHGDH inhibition: IC50 = 3 nM.
  • Disrupts serine biosynthesis in cells (IC50 ≈ 2.2 μM at 72 h).
  • Chemical formula C21H20Cl2N2O6S; molecular weight 499.36 g/mol.
  • High purity: 99.6% (reported by supplier).
  • Form: powder; suitable for DMSO stock solutions.
  • Solubility in vitro: DMSO 125 mg/mL (≈250 mM; ultrasonic recommended).
  • In vivo formulations documented for multiple vehicles at ≥2.08 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.