Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Chlorophenyl)-4,5-diphenylimidazole 98.0+%, TCI America™
SDP

Catalog No. C22405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22405G 5 g
C224025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22405G Supplier TCI America Supplier No. C22405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1707-67-1
Molecular Formula C21H15ClN2
Molecular Weight (g/mol) 330.82
MDL Number MFCD00030328
InChI Key NSWNXQGJAPQOID-UHFFFAOYSA-N
PubChem CID 74356
IUPAC Name 2-(2-chlorophenyl)-4,5-diphenyl-1H-imidazole
SMILES ClC1=CC=CC=C1C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 197°C
Color Yellow
Quantity 5 g
Formula Weight 330.82
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Chlorophenyl)-4,5-diphenylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.