Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Chlorophenyl)ethanol 97.0+%, TCI America™
SDP

Catalog No. C21691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21691G 1 g
C21695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21691G Supplier TCI America Supplier No. C21691G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19819-95-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002888
InChI Key IWNHTCBFRSCBQK-UHFFFAOYSA-N
Synonym 2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol
PubChem CID 88266
IUPAC Name 2-(2-chlorophenyl)ethanol
SMILES C1=CC=C(C(=C1)CCO)Cl

Specifications

Color Yellow
Quantity 1 g
Physical Form Clear Liquid at 20°C
Boiling Point 130°C
Formula Weight 156.61
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2-(2-Chlorophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.