Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Di-n-butyl-1,3-propanediol, 96%, Thermo Scientific™

Catalog No. AAL1561914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1561914 25 g
AAL1561906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1561914 Supplier Thermo Scientific Chemicals Supplier No. L1561914

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,2-Di-n-butyl-1,3-propanediol is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24765-57-9
Molecular Formula C11H24O2
Molecular Weight (g/mol) 188.311
MDL Number MFCD01863725
InChI Key OJMJOSRCBAXSAQ-UHFFFAOYSA-N
Synonym 2,2-di-n-butyl-1,3-propanediol, 2,2-dibutyl-1,3-propanediol, 5,5-nonanedimethanol, 2,2,-dibutylpropane-1,3-diol, 2,2-dibutyl-propane-1,3-diol, 2,2-di-n-butyl-1,3-propandiol
PubChem CID 4080430
IUPAC Name 2,2-dibutylpropane-1,3-diol
SMILES CCCCC(CCCC)(CO)CO

Specifications

Melting Point 38°C to 42°C
Boiling Point 125°C to 130°C (1 mmHg)
Quantity 25 g
Beilstein 1699746
Solubility Information Soluble in water.
Formula Weight 188.31
Percent Purity 96%
Chemical Name or Material 2,2-Di-n-butyl-1,3-propanediol
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.