Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Diallylbisphenol A

Catalog No. 413526100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-352-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-352-6100ML Supplier Sigma Aldrich Supplier No. 413526100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2G-Diallylbisphenol A (DABA) is a diallyl based epoxy modification agent that blends with the resin to improve the mechanical property of the epoxy material. It is majorly used in novalac based resins, which can be further cured with bismaleimides (BMI) for aerospace, electronics and wireless communication based applications.

Specifications

CAS 1745-89-7
Density 1.08 g/mL (at 25°C)
Molecular Formula C21H24O2
Refractive Index n20/D 1.587 (literature)
Linear Formula (CH3)2 C[C6H3(CH2CH=CH2)OH]2
MDL Number MFCD00191977
Quantity 100 mL
Synonym 2,2 -Bis(3-allyl-4-hydroxyphenyl)isopropylidene; 2,2-Bis(3-allyl-4-hydroxyphenyl)propane; 4,4 -(1-Methylethylidene)bis[2-(2-propenyl)phenol]; 4,4 -Isopropylidenebis(2-allylphenol)
Molecular Weight (g/mol) 308.41
Percent Purity 85%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.