Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Diamino-4,4'-bithiazole 98.0+%, TCI America™
SDP

Catalog No. D42731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42731G 1 g
D4273200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42731G Supplier TCI America Supplier No. D42731G

Chemical Identifiers

CAS 58139-59-6
Molecular Formula C6H6N4S2
Molecular Weight (g/mol) 198.262
MDL Number MFCD01740587
InChI Key MRFMTBTUKQIBDI-UHFFFAOYSA-N
Synonym 2,2′C-Diamino-4,4′C-bis(1,3-thiazole), 4,4′C-Bithiazole-2,2′C-diamine
PubChem CID 42654
IUPAC Name 4-(2-amino-1,3-thiazol-4-yl)-1,3-thiazol-2-amine
SMILES C1=C(N=C(S1)N)C2=CSC(=N2)N

Specifications

Melting Point 250°C
Quantity 1 g
Formula Weight 198.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Diamino-4,4′-bithiazole
RTECSNumber EB9000000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.