Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dichloroacetophenone 97.0+%, TCI America™
SDP

Catalog No. D112225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D112225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D112225G Supplier TCI America Supplier No. D112225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2648-61-5
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00000844
InChI Key CERJZAHSUZVMCH-UHFFFAOYSA-N
Synonym 2,2-dichloroacetophenone, phenacylidene chloride, alpha,alpha-dichloroacetophenone, 1,1-dichloroacetophenone, ethanone, 2,2-dichloro-1-phenyl, acetophenone, 2,2-dichloro, unii-3vj32jj8lh, .alpha.,.alpha.-dichloroacetophenone, 3vj32jj8lh, 2,2-dichloro-1-phenyl-ethanone
PubChem CID 72870
IUPAC Name 2,2-dichloro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C(Cl)Cl

Specifications

Boiling Point 247°C
Quantity 25 g
Formula Weight 189.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dichloroacetophenone
RTECSNumber AM7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.