Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,2-Dichlorocyclopropyl)benzene 98.0+%, TCI America™
SDP

Catalog No. D44105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44105G 5 g
D441025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44105G Supplier TCI America Supplier No. D44105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2415-80-7
Molecular Formula C9H8Cl2
Molecular Weight (g/mol) 187.063
MDL Number MFCD00001272
InChI Key WLWFQGXZIDYWQF-UHFFFAOYSA-N
Synonym 2,2-dichlorocyclopropyl benzene, 1,1-dichloro-2-phenylcyclopropane, 2,2-dichlorocyclopropyl-benzene, benzene, 2,2-dichlorocyclopropyl, cyclopropane, 1,1-dichloro, 2-phenyl, pubchem18679, acmc-1clka, ksc491m2n, wlwfqgxzidywqf-uhfffaoysa, 2,2-dichloro-cyclopropyl-benzene
PubChem CID 95349
IUPAC Name (2,2-dichlorocyclopropyl)benzene
SMILES C1C(C1(Cl)Cl)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 187.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (2,2-Dichlorocyclopropyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.