Learn More
Strem, An Ascensus Company CAS# 1067883-58-2. 100mg. 2-[2-(Dicyclohexylphosphino)phenyl]-1-methyl-1H-indole, min. 98% CM-Phos. MFCD16995285
SDP
Supplier:  Strem, An Ascensus Company 151088100MG
Encompass
CAS# 1067883-58-2. 100mg. 2-[2-(Dicyclohexylphosphino)phenyl]-1-methyl-1H-indole, min. 98% CM-Phos. MFCD16995285. Molecular Weight: 403.54. Molecular Formula: C27H34NP. Color/form: white to off-white pwdr. Strem# 15-1088. http://www.strem.com/catalog/v/15-1088/
Catalog No. 50-814-83