Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2-Difluoroacetophenone

Catalog No. 7938411G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-384-11G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 79-384-11G Supplier Sigma Aldrich Supplier No. 7938411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Product was reported by Hartwig and coworkers in the one-pot synthesis of difluoromethylated arenes from aryl chlorides and bromides.Pd-Catalyzed a-Arylation of a;a-Difluoroketones with Aryl Bromides and Chlorides. A Route to Difluoromethylarenes

Specifications

CAS 138048-80-3
Boiling Point 128°C to 129°C (13 mmHg, lit.)
Molecular Formula C29H33NO
Linear Formula C29H33NO
MDL Number MFCD01867502
Quantity 25 g
Molecular Weight (g/mol) 411.58 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.