Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2-Difluorosuccinic acid

Catalog No. CDS002643 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002643 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002643 Supplier Sigma Aldrich Supplier No. CDS00264350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-chloro-1,4-benzoquinone is a quinone derivative
  • It is one of the intermediate formed during the degradation of 3,4-dichloroaniline in a dielectric barrier discharge plasma reactor
  • It is formed during lignin peroxidase catalyzed oxidation of 2-chloro-1,4-dimethoxybenzene
  • 2-chloro-1,4-benzoquinone on dechlorination yields 1,2,4-trihydroxybenzene.2-chloro-1,4-benzoquinone may be used in the preparation of chloro derivatives of prenylnaphthohydroquinone

Specifications

CAS 665-31-6
Molecular Formula C4H4F2O4
Linear Formula C4H4F2O4
MDL Number MFCD00040970
Quantity 5 g
Molecular Weight (g/mol) 154.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.