Learn More
Medchemexpress LLC 2,2-Dihydroxy-1-phenylethan-1-one | 1075-06-5 | 97.0% | 25 G
SDP
Catalog No. 5000341783
Encompass_Preferred
2,2-Dihydroxy-1-phenylethan-1-one is an arylglyoxal monohydrate that serves as an intermediate in pharmaceutical synthesis. It possesses antioxidant properties and can be utilized as a starting material for the synthesis of aromatic or aliphatic amines.
  • An arylglyoxal monohydrate.
  • Intermediate for pharmaceutical synthesis.
  • Exhibits antioxidant property.
  • Starting material for synthesis of aromatic or aliphatic amines.
Catalog No. 50-003-41783 Supplier Medchemexpress LLC Supplier No. 25GHY128407
2,2-Dihydroxy-1-phenylethan-1-one is an arylglyoxal monohydrate that serves as an intermediate in pharmaceutical synthesis. It possesses antioxidant properties and can be utilized as a starting material for the synthesis of aromatic or aliphatic amines.
  • An arylglyoxal monohydrate.
  • Intermediate for pharmaceutical synthesis.
  • Exhibits antioxidant property.
  • Starting material for synthesis of aromatic or aliphatic amines.