Learn More
Medchemexpress LLC 2,2-Dihydroxy-1-phenylethan-1-one | 1075-06-5 | 97.0% | 5 G
SDP
Catalog No. 5000339050
Encompass_Preferred
2,2-Dihydroxy-1-phenylethan-1-one, also known as Phenylglyoxal hydrate, is an arylglyoxal monohydrate. It is an intermediate used in pharmaceutical synthesis and exhibits antioxidant properties. This compound serves as a starting material for the synthesis of aromatic or aliphatic amines.
  • Intermediate in pharmaceutical synthesis
  • Possesses antioxidant properties
  • Can be used as a starting material for synthesizing aromatic or aliphatic amines
Catalog No. 50-003-39050 Supplier Medchemexpress LLC Supplier No. HY1284075G
2,2-Dihydroxy-1-phenylethan-1-one, also known as Phenylglyoxal hydrate, is an arylglyoxal monohydrate. It is an intermediate used in pharmaceutical synthesis and exhibits antioxidant properties. This compound serves as a starting material for the synthesis of aromatic or aliphatic amines.
  • Intermediate in pharmaceutical synthesis
  • Possesses antioxidant properties
  • Can be used as a starting material for synthesizing aromatic or aliphatic amines