Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dihydroxy-4,4'-dimethoxybenzophenone 90.0+%, TCI America™
SDP

Catalog No. D057525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D057525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D057525G Supplier TCI America Supplier No. D057525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131-54-4
Molecular Formula C15H14O5
Molecular Weight (g/mol) 274.272
MDL Number MFCD00009606
InChI Key SODJJEXAWOSSON-UHFFFAOYSA-N
Synonym 2,2'-dihydroxy-4,4'-dimethoxybenzophenone, bis 2-hydroxy-4-methoxyphenyl methanone, benzophenone-6, cyasorb uv 12, methanone, bis 2-hydroxy-4-methoxyphenyl, uvinul d 49, caswell no. 353c, unii-7813j9cs1g, 2,2-dihydroxy-4,4-dimethoxybenzophenone, 4,4'-dimethoxy-2,2'-dihydroxybenzophenone
PubChem CID 8570
IUPAC Name bis(2-hydroxy-4-methoxyphenyl)methanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=C(C=C(C=C2)OC)O)O

Specifications

Melting Point 135°C
Quantity 25 g
Formula Weight 274.27
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dihydroxy-4,4′-dimethoxybenzophenone
EINECSNumber (3)-1261&(4)-0158
RTECSNumber DJ0900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.