Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Dihydroxy-4-methoxybenzophenone

Catalog No. 32357825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-357-825G 25 g
32-357-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-357-825G Supplier Sigma Aldrich Supplier No. 32357825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2'-Dihydroxy-4-methoxybenzophenone

Specifications

CAS 131-53-3
Melting Point 73°C to 75°C
Boiling Point 170°C to 175°C (1 mmHg)
Molecular Formula C14H12O4
Linear Formula CH3OC6H3(OH)COC6H4OH
Quantity 25 g
MDL Number MFCD00002218
Synonym Dioxybenzone
Molecular Weight (g/mol) 244.24
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.