Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dihydroxy-4-methoxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. D058625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D058625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D058625G Supplier TCI America Supplier No. D058625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 131-53-3
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00002218
InChI Key MEZZCSHVIGVWFI-UHFFFAOYSA-N
Synonym dioxybenzone, 2,2'-dihydroxy-4-methoxybenzophenone, dioxybenzon, benzophenone-8, advastab 47, cyasorb uv 24, spectra-sorb uv 24, 2-hydroxy-4-methoxyphenyl 2-hydroxyphenyl methanone, methanone, 2-hydroxy-4-methoxyphenyl 2-hydroxyphenyl, cyasorb uv 24 light absorber
PubChem CID 8569
ChEBI CHEBI:34208
IUPAC Name (2-hydroxy-4-methoxyphenyl)-(2-hydroxyphenyl)methanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2O)O

Specifications

Melting Point 71°C
Color Yellow
Boiling Point 175°C
Quantity 25 g
Formula Weight 244.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dihydroxy-4-methoxybenzophenone
EINECSNumber (4)-0149
RTECSNumber DJ1049500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.