Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,2-dihydroxyacetic acid | 563-96-2 | MFCD00127974 | 98.0% | 92.05 g/mol | C2H4O4 | 50 G
SDP

Catalog No. 5000299550
Encompass_Preferred

2,2-Dihydroxyacetic acid is a small, research-grade carboxylic acid used as an intermediate in synthesis and laboratory research. It is supplied as a solid with a molecular weight of 92.05 g/mol and reported purity suitable for general research applications.

  • High reported purity of 98.0% for research use.
  • Supplied as a solid, white to light yellow, convenient for storage and handling.
  • Molecular formula C2H4O4 and molecular weight 92.05 g/mol for precise formulation.
  • Suitable as a chemical intermediate in organic synthesis.
  • SMILES string available for cheminformatics applications.

Catalog No. 50-002-99550 Supplier Medchemexpress LLC Supplier No. HYW01972450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,2-Dihydroxyacetic acid is a small, research-grade carboxylic acid used as an intermediate in synthesis and laboratory research. It is supplied as a solid with a molecular weight of 92.05 g/mol and reported purity suitable for general research applications.

  • High reported purity of 98.0% for research use.
  • Supplied as a solid, white to light yellow, convenient for storage and handling.
  • Molecular formula C2H4O4 and molecular weight 92.05 g/mol for precise formulation.
  • Suitable as a chemical intermediate in organic synthesis.
  • SMILES string available for cheminformatics applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.