Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dihydroxydiphenylmethane 99.0+%, TCI America™
SDP

Catalog No. D19395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19395G 5 g
D19391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19395G Supplier TCI America Supplier No. D19395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2467-02-9
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00002243
InChI Key MQCPOLNSJCWPGT-UHFFFAOYSA-N
Synonym 2,2'-methylenediphenol, bis 2-hydroxyphenyl methane, 2,2'-dihydroxydiphenylmethane, 2,2'-bisphenol f, 2,2'-methylenebis phenol, phenol, 2,2'-methylenebis, o-o-hydroxybenzyl phenol, 2-2-hydroxyphenyl methyl phenol, phenol, 2,2'-methylenedi, 2,2'-bis hydroxyphenyl methane
PubChem CID 75575
ChEBI CHEBI:34207
IUPAC Name 2-[(2-hydroxyphenyl)methyl]phenol
SMILES C1=CC=C(C(=C1)CC2=CC=CC=C2O)O

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Formula Weight 200.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dihydroxydiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.