Learn More
Medchemexpress LLC 2 2-Dimethoxy-2-phen 100g
SDP
Catalog No. 5000204617
Encompass_Preferred
2 2-Dimethoxy-2-phenylacetophenone (Photoinitiator-651) is a photoinitiator that by absorbing UV light generates free radicals to initiate cross-linking reactions thereby curing coating materials and forming adhesive coatings 2 2-Dimethoxy-2-phenylacetophenone can be used in research on the preparation of conductive adhesives 3D printing materials medical coatings and biocompatible glues for biomedical devices[1]
Catalog No. 50-002-04617 Supplier Medchemexpress LLC Supplier No. HYW098021100G
2 2-Dimethoxy-2-phenylacetophenone (Photoinitiator-651) is a photoinitiator that by absorbing UV light generates free radicals to initiate cross-linking reactions thereby curing coating materials and forming adhesive coatings 2 2-Dimethoxy-2-phenylacetophenone can be used in research on the preparation of conductive adhesives 3D printing materials medical coatings and biocompatible glues for biomedical devices[1]