Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2-Dimethoxypropane

Catalog No. D13680825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13680825ML 25 mL
D13680818L 18 L
D13680825L 2.5 L
D136808 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. D13680825ML Supplier Sigma Aldrich Supplier No. D13680825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2-Dimethoxypropane (DMP) is an organic building block commonly employed as a precursor to generate 2-methoxypropene (MPP). The degradation study of DMP in ionic liquids showed the formation of MPP and 2-ethoxypropene (EPP) in an identical ratio due to the tunneling effect. Conformational analysis of DMP based on ab initio calculations and matrix isolation infrared spectroscopy has been reported. DMP reacts with water to produce methanol and acetone. This reaction has been employed in a method for the quantification of water in natural products by gas-liquid chromatography. Acidified DMP has been employed for the dehydration of biological samples.Reagent for the preparation of 1,2-diols as acetonides.

Specifications

CAS 77-76-9
Density 0.847 g/mL at 25°C
Boiling Point 83°C
Molecular Formula C5H12O2
Refractive Index n20/D 1.378
Linear Formula (CH3)2C(OCH3)2
MDL Number MFCD00008479
Quantity 25 mL
Synonym Acetone dimethyl acetal
Molecular Weight (g/mol) 104.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.