Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethoxypropane, ≥99.4% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019099 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9099 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9099 Supplier MilliporeSigma Supelco Supplier No. 731375MLF
Add to Cart
Add to Cart

2,2-Dimethoxypropane is an organic compound, which may be used as a protecting agent in the process of the synthesis of structurally related alkaloids like narciclasine and lycoricidine using phenylbromide as the starting material.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 77-76-9
Vapor Pressure 60 mmHg (15.8°C)
Vapor Density 3.59 (vs. air)
Density 0.847 g/mL (at 25°C)
Flash Point -10°C
Physical Form Neat
Linear Formula (CH3)2 C(OCH3)2
MDL Number MFCD00008479
Boiling Point 79°C to 81°C; 83°C
Synonym Acetone dimethyl acetal
Type Analytical Standard
Quantity 5 mL
Formula Weight 104.15
Percent Purity ≥99.4% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.