Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2-Dimethyl-1,3-dioxane-4,6-dione

Catalog No. 210145100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-014-5100G 100 g
21-014-510G 10 g
21-014-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-014-5100G Supplier Sigma Aldrich Supplier No. 210145100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2-Dimethyl-1,3-dioxane-4,6-dione

Specifications

CAS 2033-24-1
Melting Point 92°C to 96°C (lit.)
Molecular Formula C6H8O4
Linear Formula C6H8O4
MDL Number MFCD00006638
Quantity 100 g
Synonym Malonic acid cyclic isopropylidene ester; Meldrums acid; cycl-Isopropylidene malonate
Molecular Weight (g/mol) 144.13
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.