Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-1,3-dioxolane 99%, Thermo Scientific™

Catalog No. AC338250050
Change view
Click to view available options
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC338250050 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338250050 Supplier Thermo Scientific Chemicals Supplier No. 338250050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2916-31-6
Density 0.95
Infrared Spectrum Authentic
Molecular Formula C5H10O2
Refractive Index 1.3970 to 1.3990 (20°C, 589nm)
MDL Number MFCD00014106
Quantity 5mL
Synonym 1,3-dioxolane, 2,2-dimethyl, 2,2-dimethyl-1,3-dioxolan, 1,3-dioxolane, dimethyl, acetone ethylene ketal, acetone ethylene acetal, acetone, cyclic ethylene acetal, 1,3-dioxolane,2,2-dimethyl, sijbdwpvnayvgy-uhfffaoysa, 2,2-dimethyl-1,3-dioxolane
InChI Key SIJBDWPVNAYVGY-UHFFFAOYSA-N
SMILES CC1(C)OCCO1
IUPAC Name 2,2-dimethyl-1,3-dioxolane
Molecular Weight (g/mol) 102.13
PubChem CID 76209
Formula Weight 102.13
Percent Purity ≥98.5%
Chemical Name or Material 2, 2-Dimethyl-1, 3-dioxolane, 99%
Show More Show Less
EINECSNumber 220-842-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.