Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2-Dimethyl-1,3-propanediol

Catalog No. 538256500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-825-6500G 500 g
53-825-63KG 3 kg
53-825-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 53-825-6500G Supplier Sigma Aldrich Supplier No. 538256500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,2-dimethyl-1,3-propanediol readily absorbs moisture on exposure to air.2,2-dimethyl-1,3-propanediol may be used in the synthesis of:
  • 2,2-dimethyl-1,3-propanediol cyclic phosphorochloridate
  • 2,2-dimethyl-1,3-propanediol cyclic phenylphosphonate
  • [1α,3α-(2α,2α-dimethylpropylene)]-2-iodo-3-octyl-5-thienylboronate
  • Cyclic carbonate
  • 2,2-dimethyl-1,3-propanediol bis(cyclic-2,2-dimethyltrimethylene phosphite)

Specifications

CAS 126-30-7
Molecular Formula C5H12O2
Linear Formula (CH3)2 C(CH2OH)2
MDL Number MFCD00004685
Quantity 500 g
Synonym NPG Glycol; Neopentylglycol
Molecular Weight (g/mol) 104.15
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.