Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-3-(4-methylpiperidino)propanal, 97%, Thermo Scientific™

Catalog No. MO01200DA
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MO01200DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01200DA Supplier Thermo Scientific Chemicals Supplier No. MO01200DA

missing translation for 'validMainframeMsg'

Specifications

CAS 690632-29-2
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C11H21NO
Quantity 1g
MDL Number MFCD05022484
Synonym 2,2-dimethyl-3-4-methylpiperidin-1-yl propanal, 2,2-dimethyl-3-4-methyl-piperidin-1-yl-propionaldehyde, 2,2-dimethyl-3-4-methylpiperidino propanal, 2,2-dimethyl-3-4-methylpiperidyl propanal
InChI Key RLPZZDZTGNLJEH-UHFFFAOYSA-N
SMILES CC1CCN(CC(C)(C)C=O)CC1
IUPAC Name 2,2-dimethyl-3-(4-methylpiperidin-1-yl)propanal
Molecular Weight (g/mol) 183.30
PubChem CID 2794707
Formula Weight 183.29
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-3-(4-methylpiperidino)propanal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.