Learn More
Apexbio Technology LLC 2-2-Dimethyl Succinic acid 597-43-3 500mg
SDP
Catalog No. 5000092390
Encompass_Preferred
2 2-Dimethyl succinic acid is a succinate derivative characterized by structural alkyl substitution It functions through interaction with mitochondrial metabolic pathways particularly by interfering with enzymatic activities within the tricarboxylic acid (TCA) cycle thereby affecting mitochondrial function and intracellular redox balance 2 2-Dimethyl succinic acid exerts its biological activity primarily through modulation of mitochondrial bioenergetics via disruption of TCA cycle enzymatic processes Based on these properties 2 2-dimethyl succinic acid holds research potential as a biological intermediate or reference compound in biomedical studies of mitochondrial dysfunction metabolic regulation mechanisms cancer neurodegenerative diseases and metabolic disorders
Catalog No. 50-000-92390 Supplier Apexbio Technology LLC Supplier No. B7856500
2 2-Dimethyl succinic acid is a succinate derivative characterized by structural alkyl substitution It functions through interaction with mitochondrial metabolic pathways particularly by interfering with enzymatic activities within the tricarboxylic acid (TCA) cycle thereby affecting mitochondrial function and intracellular redox balance 2 2-Dimethyl succinic acid exerts its biological activity primarily through modulation of mitochondrial bioenergetics via disruption of TCA cycle enzymatic processes Based on these properties 2 2-dimethyl succinic acid holds research potential as a biological intermediate or reference compound in biomedical studies of mitochondrial dysfunction metabolic regulation mechanisms cancer neurodegenerative diseases and metabolic disorders