Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals

Catalog No. AAL1466403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1466403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1466403 Supplier Thermo Scientific Chemicals Supplier No. L1466403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 813-72-9 | Molecular Formula: C8H16O2 | Molecular Weight: 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-72-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00965097
InChI Key YTTWDTVYXAEAJA-UHFFFAOYSA-N
Synonym 2,2-dimethyl-hexanoic acid, hexanoic acid,2,2-dimethyl, 2,2-dimethylhexanoicacid, dimethylhexanoic acid, dimethyl-hexanoic acid, acmc-209plb, 2,2-dimethyl hexanoic acid, #, hexanoic acid, 2,2-dimethyl, yttwdtvyxaeaja-uhfffaoysa
PubChem CID 163217
IUPAC Name 2,2-dimethylhexanoic acid
SMILES CCCCC(C)(C)C(O)=O

Specifications

Density 0.913
Boiling Point 216°C to 220°C
Refractive Index 1.427
Quantity 1 g
UN Number UN3265
Beilstein 1749961
Formula Weight 144.22
Percent Purity 94%
Chemical Name or Material 2,2-Dimethylhexanoic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.