Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylvaleronitrile 98.0+%, TCI America™
SDP

Catalog No. D41715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41715G Supplier TCI America Supplier No. D41715G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20654-47-1
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.188
InChI Key TZJQCUDHKUWEFU-UHFFFAOYSA-N
Synonym 2,2-Dimethylpentanenitrile
PubChem CID 12754185
IUPAC Name 2,2-dimethylpentanenitrile
SMILES CCCC(C)(C)C#N

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 g
UN Number 1993
Formula Weight 111.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylvaleronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.