Learn More
Medchemexpress LLC 2,2-diphenylacetonitrile | 86-29-3 | MFCD00001862 | 99.7% | 193.24 g/mol | C14H11N | 25 G
SDP
Catalog No. 5000303565
Encompass_Preferred
2,2-Diphenylacetonitrile (CAS 86-29-3) is an organic reagent used as a synthetic building block and intermediate in organic and life-science research. It is supplied for laboratory research use only and is characterized by high purity and well-defined physical properties suitable for analytical and synthetic applications.
  • High purity (~99.7%) suitable for synthesis and analysis.
  • Molecular formula C14H11N; molecular weight 193.24 g/mol.
  • Solid crystalline material with a melting point around 73-76 °C.
  • Available in multiple laboratory-scale sizes for flexible use.
  • For research use only; not for human or clinical applications.
Catalog No. 50-003-03565 Supplier Medchemexpress LLC Supplier No. 25GHYW015052
2,2-Diphenylacetonitrile (CAS 86-29-3) is an organic reagent used as a synthetic building block and intermediate in organic and life-science research. It is supplied for laboratory research use only and is characterized by high purity and well-defined physical properties suitable for analytical and synthetic applications.
  • High purity (~99.7%) suitable for synthesis and analysis.
  • Molecular formula C14H11N; molecular weight 193.24 g/mol.
  • Solid crystalline material with a melting point around 73-76 °C.
  • Available in multiple laboratory-scale sizes for flexible use.
  • For research use only; not for human or clinical applications.