Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diphenylethylamine 98.0+%, TCI America™
SDP

Catalog No. D20185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20185G 5 g
D201825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20185G Supplier TCI America Supplier No. D20185G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3963-62-0
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00008143
InChI Key RXMTUVIKZRXSSM-UHFFFAOYSA-N
Synonym 2,2-diphenylethylamine, 2,2-diphenyl-ethylamine, 1-amino-2,2-diphenylethane, 2,2-diphenylethan-1-amine, chembl10780, pubchem7986, acmc-1cswz, 2,2-diphenylethyl-amine, 2,2-di phenyl ethanamine
PubChem CID 77575
IUPAC Name 2,2-diphenylethan-1-amine
SMILES NCC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 49°C
Quantity 5 g
Formula Weight 197.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Diphenylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.