Learn More
Medchemexpress LLC 2-(2-(Diphenylphosphino)ethyl)pyridine | 10150-27-3 | 291.33 | 250 MG
SDP
Catalog No. 5000453278
Encompass_Preferred
2-(2-(Diphenylphosphino)ethyl)pyridine is an important chiral catalyst with excellent catalytic activity. It is widely used in asymmetric synthesis reactions, especially in the field of compound synthesis. It can also be used in metal-catalyzed reactions to improve the selectivity and yield of the reaction.
  • Important chiral catalyst with excellent catalytic activity.
  • Widely used in asymmetric synthesis reactions.
  • Can be used in metal-catalyzed reactions to improve selectivity and yield.
Catalog No. 50-004-53278 Supplier Medchemexpress LLC Supplier No. 250MGHYW009283
Add to Cart
Add to Cart
2-(2-(Diphenylphosphino)ethyl)pyridine is an important chiral catalyst with excellent catalytic activity. It is widely used in asymmetric synthesis reactions, especially in the field of compound synthesis. It can also be used in metal-catalyzed reactions to improve the selectivity and yield of the reaction.
  • Important chiral catalyst with excellent catalytic activity.
  • Widely used in asymmetric synthesis reactions.
  • Can be used in metal-catalyzed reactions to improve selectivity and yield.